missing translation for 'onlineSavingsMsg'
Learn More

Sodium fumarate, 98%, Thermo Scientific Chemicals

Product Code. 10772505
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17013-01-3
Molecular Formula C4H2Na2O4
Molecular Weight (g/mol) 160.04
MDL Number MFCD00064567
InChI Key VXXVUHAXJHEYFH-SEPHDYHBSA-N
Synonym disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate
PubChem CID 87110070
IUPAC Name (E)-but-2-enedioic acid;sodium
SMILES C(=CC(=O)O)C(=O)O.[Na].[Na]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NaO2CCH=CHCO2Na
Quantity 500 g
Beilstein 02,74
Merck Index 10,4165
Solubility Information Solubility in water: 228g/L water. Other solubilities: insoluble in ethanol, aceton
Formula Weight 160.04
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Sodium fumarate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'