missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Reduction reagent
Specifications
Specifications
| Melting Point | 800.0°C |
| Color | Gray to Yellow |
| Physical Form | Tacky Powder |
| Quantity | 500 g |
| Linear Formula | NaH |
| Fieser | 02,382; 01,1075; 04,452; 05,610; 06,541; 07,335; 08,458; 09,427; 11,486; 12,447; 14,288; 16,307 |
| Flash Point | 165°C |
| Merck Index | 15,876 |
| Solubility Information | Solubility in water: reacts. Other solubilities: reacts with alcohols, soluble in molten sodium hydroxide, insoluble in cs2, ccl4, liquid nh3, benzene |
| Formula Weight | 24 |
| Show More |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur