Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals
Product Code.: 12307103
Product Code. 12307103
2 kg
Click to view available options
Quantity:
100 g
500 g
2 kg
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code.
12307103
Brand: Thermo Scientific Acros
332140020
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Reduction reagent
Chemical Identifiers
CAS
7646-69-7, 8042-47-5
Molecular Formula
HNa
Molecular Weight (g/mol)
24
MDL Number
MFCD00003471
InChI Key
BZKBCQXYZZXSCO-UHFFFAOYSA-N
Synonym
sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydrid Show More Show Less
PubChem CID
24758
IUPAC Name
sodium;hydride
SMILES
[H-].[Na+]
Specifications
Melting Point
800.0°C
Color
Gray to Yellow
Physical Form
Tacky Powder
Quantity
2 kg
Linear Formula
NaH
Fieser
02,382; 01,1075; 04,452; 05,610; 06,541; 07,335; 08,458; 09,427; 11,486; 12,447; 14,288; 16,307
Flash Point
165°C
Merck Index
15,876
Solubility Information
Solubility in water: reacts. Other solubilities: reacts with alcohols, soluble in molten sodium hydroxide, insoluble in cs2, ccl4, liquid nh3, benzene
Formula Weight
24
Percent Purity
57 to 63% (NaH)
Packaging
Metal Can
Density
1.2000g/mL
Chemical Name or Material
Sodium hydride, 60% dispersion in mineral oil, In soluble bags, in resealable cans
Show More
Show Less
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number
Certificate Type
Certificate of Analysis
Certificate of Compliance
Certificate of Sterility/Irradiation
Certificate of Validation
Certificate of Guarantee
Certificate of Animal Origin
Certificate of Quality
Certificate of Processing
Declaration of Conformity
Search
5 results found
Lot Number
Certificate Type
Date
Catalog Number
Lot Number A0402180
Certificate Type Certificate of Analysis
Date 14/12/2022
Catalog Number 332141000,332140020,332145000
Lot Number A0395478
Certificate Type Certificate of Analysis
Date 14/12/2022
Catalog Number 332141000,332140020,332145000
Lot Number A0403795
Certificate Type Certificate of Analysis
Date 14/12/2022
Catalog Number 332141000,332140020,332145000
Lot Number A0396618
Certificate Type Certificate of Analysis
Date 14/12/2022
Catalog Number 332141000,332140020,332145000
Lot Number A0395477
Certificate Type Certificate of Analysis
Date 14/12/2022
Catalog Number 332141000,332140020,332145000
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.