missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydride, 60% dispersion in mineral oil, in soluble bags, Thermo Scientific Chemicals

36.52 € - 125.00 €

Chemical Identifiers

CAS 7646-69-7, 8042-47-5
Molecular Formula HNa
Molecular Weight (g/mol) 24
MDL Number MFCD00003471
InChI Key BZKBCQXYZZXSCO-UHFFFAOYSA-N
Synonym sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydrid
PubChem CID 24758
IUPAC Name sodium;hydride
SMILES [H-].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10104210
SDS View Documents
Thermo Scientific Acros
189861000
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10646772
SDS View Documents
Thermo Scientific Acros
189862500
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10367270
SDS View Documents
Thermo Scientific Acros
189860010
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Reduction reagent
Specifications

Chemical Identifiers

7646-69-7, 8042-47-5
24
BZKBCQXYZZXSCO-UHFFFAOYSA-N
24758
[H-].[Na+]
HNa
MFCD00003471
sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydrid
sodium;hydride

Specifications

800.0°C
Gray to Yellow
HNa
MFCD00003471
165°C
sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydrid
BZKBCQXYZZXSCO-UHFFFAOYSA-N
sodium;hydride
24758
57 to 63% (NaH)
Sodium hydride, 60% dispersion in mineral oil
7646-69-7, 8042-47-5
Tacky Powder
NaH
02,382; 01,1075; 04,452; 05,610; 06,541; 07,335; 08,458; 09,427; 11,486; 12,447; 14,288; 16,307
15, 8760
Solubility in water: reacts. Other solubilities: reacts with alcohols, soluble in molten sodium hydroxide, insoluble in cs2, ccl4, liquid nh3, benzene
[H-].[Na+]
24
24
1.2000g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement:


WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Product Identifier
  • Sodium hydride, 60% dispersion in mineral oil

Signal Word
  • Danger

Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.

Supplemental information
  • MIXTURE LIST-Contains : Sodium hydride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.