missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydrogen arsenate heptahydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Sodium hydrogen arsenate heptahydrate is used as a source of soluble arsenic and in dyeing and printing with Turkey-red oil. It also acts as a veterinary drug for anti-parasitic, skin diseases, blood diseases, rheumatism, and asthma.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water and glycerol.
Notes
Incompatible with strong oxidizing agents and strong acids.
Soluble in water and glycerol.
Notes
Incompatible with strong oxidizing agents and strong acids.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 130°C (decomposition) |
Quantity | 500 g |
UN Number | UN1685 |
Solubility Information | Soluble in water and glycerol. |
Formula Weight | 312.01 (185.94 Anhydrous) |
Percent Purity | 98% |
Odor | Odorless |
Density | 1.88 |
Chemical Name or Material | Sodium hydrogen arsenate heptahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'