missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydrogen Carbonate, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 500g, CHEBI:32139, White, 84.01g/mol, UIIMBOGNXHQVGW-UHFFFAOYSA-M
Chemical Identifiers
CAS | 144-55-8 |
---|---|
Molecular Formula | CHNaO3 |
Molecular Weight (g/mol) | 84.01 |
MDL Number | MFCD00003528 |
InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
Synonym | sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut |
PubChem CID | 516892 |
ChEBI | CHEBI:32139 |
SMILES | [Na+].OC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10020510
|
Fisher Chemical
S/4240/53 |
500 g | Plastic powder jar |
N/A
|
N/A
|
||||
10725921
|
Fisher Chemical
S/4240/63 |
3 kg | Plastic powder jar |
N/A
|
N/A | ||||
10518640
|
Fisher Chemical
S/4240/65 |
5 kg | Plastic keg |
N/A
|
N/A | ||||
Description
CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 500g, CHEBI:32139, White, 84.01g/mol, UIIMBOGNXHQVGW-UHFFFAOYSA-M
Chemical Identifiers
144-55-8 | |
84.01 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
516892 | |
[Na+].OC([O-])=O |
CHNaO3 | |
MFCD00003528 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
CHEBI:32139 |
Specifications
270°C | |
White | |
Powder | |
MFCD00003528 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
sodium;hydrogen carbonate | |
516892 | |
84.01g/mol |
144-55-8 | |
8.3 | |
CHNaO3 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
[Na+].OC([O-])=O | |
84.01 | |
CHEBI:32139 |
Safety and Handling
Product Identifier
- Sodium bicarbonate