Learn More
Sodium Hydrogen Carbonate, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 1kg, CHEBI:32139, White, 84.01g/mol, UIIMBOGNXHQVGW-UHFFFAOYSA-M
Brand: Fisher Chemical S/4240/60
34.01 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Description
CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 1kg, CHEBI:32139, White, 84.01g/mol, UIIMBOGNXHQVGW-UHFFFAOYSA-M
Specifications
270°C | |
White | |
Powder | |
CHNaO3 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
[Na+].OC([O-])=O | |
84.01 | |
CHEBI:32139 | |
Plastic powder jar |
144-55-8 | |
8.3 | |
1 kg | |
MFCD00003528 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
sodium;hydrogen carbonate | |
516892 | |
84.01g/mol |
Safety and Handling
- Sodium bicarbonate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.