missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrogen Carbonate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10122640
Click to view available options
Quantity:
1 kg
3 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar

Sodium Hydrogen Carbonate, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270°C
Color White
pH 8.3
Physical Form Powder/Solid
Quantity 5 kg
IUPAC Name sodium;hydrogen carbonate
Formula Weight 84.01g/mol
Packaging Plastic keg
Product Identifier
  • Sodium bicarbonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'