Learn More
Sodium hydrogen carbonate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
m.p. approx. 50°C, begins to lose CO2 approx.100°C, converts to Na2CO3
166.83 € - 607.02 €
Chemical Identifiers
CAS | 144-55-8 |
---|---|
Molecular Formula | CHNaO3 |
Molecular Weight (g/mol) | 84.01 |
MDL Number | MFCD00003528 |
InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
Synonym | sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut |
PubChem CID | 516892 |
ChEBI | CHEBI:32139 |
SMILES | [Na+].OC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317027
|
Thermo Scientific Alfa Aesar
010863.09 |
10 g |
166.83 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307027
|
Thermo Scientific Alfa Aesar
010863.18 |
50 g |
607.02 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium hydrogen carbonate is used in a process for removing paint and corrosion called sodablasting; the process is particularly suitable for cleaning aluminum panels which can be distorted by other types of abrasive. It can be administered to pools, spas, and garden ponds to raise pH levels. Also can be used as a fire extinguisher. Its primary use is in cooking (baking), as a leavening agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
144-55-8 | |
84.01 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
516892 | |
[Na+].OC([O-])=O |
CHNaO3 | |
MFCD00003528 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
CHEBI:32139 |
Specifications
144-55-8 | |
Powder | |
CHNaO3 | |
4153970 | |
Soluble in water. Insoluble in ethanol. | |
[Na+].OC([O-])=O | |
84.01 | |
CHEBI:32139 | |
Odorless | |
White powder | |
Sodium hydrogen carbonate |
White | |
99.998% (metals basis) | |
MFCD00003528 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
sodium hydrogen carbonate | |
516892 | |
84.01 | |
1.5 | |
2.159 |
Safety and Handling
H303: May be harmful if swallowed.
missing translation for 'einecsNumber' : 205-633-8
missing translation for 'rtecsNumber' : VZ0950000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.