missing translation for 'onlineSavingsMsg'
Learn More
Sodium hydrogen L-tartrate, anhydrous, 98%, Thermo Scientific Chemicals Product Code.: 11313176

Sodium hydrogen L-tartrate, anhydrous, 98%, Thermo Scientific Chemicals

Product Code. 11313176
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Product Code. 11313176

Brand: Thermo Scientific Alfa Aesar L12469.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 526-94-3
Molecular Formula C4H5NaO6
Molecular Weight (g/mol) 172.07
MDL Number MFCD00065393,MFCD00065393
InChI Key NKAAEMMYHLFEFN-UHFFFAOYNA-M
Synonym sodium bitartrate, monosodium tartrate, sodium hydrogen tartrate, natriumtartrat german, sodium 3-carboxy-2,3-dihydroxypropanoate, monobasic sodium tartrate, monosodium l-+-tartrate, tartaric acid, monosodium salt, natrium hydrogen-2r,3r-tartrat, weinsteinShow MoreShow Less
PubChem CID 23690454
IUPAC Name sodium;2,3,4-trihydroxy-4-oxobutanoate
SMILES [Na+].OC(C(O)C([O-])=O)C(O)=O

Specifications

Melting Point ∽243°C (decomposition)
Quantity 25 g
Beilstein 6099125
Merck Index 14,8589
Solubility Information Soluble in water.
Optical Rotation +24.5° (c=3 in Water)
Formula Weight 172.07
Percent Purity 98%
Chemical Name or Material Sodium hydrogen L-tartrate, anhydrous

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241716 Certificate TypeCertificate of Analysis Date07/07/2022 Catalog NumberL12469.22,L12469.14,L12469.36
Lot Number10108061 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL12469.22,L12469.14,L12469.36
Lot Number10152794 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL12469.22,L12469.14,L12469.36
Lot Number10176644 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL12469.22,L12469.14,L12469.36
Lot Number10152097 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL12469.22,L12469.14,L12469.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium hydrogen L-tartrate, anhydrous, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.