Learn More
Sodium hydrogen phosphate, anhydrous, ACS, 99.0% min, Thermo Scientific Chemicals
25.43 € - 236.18 €
Chemical Identifiers
CAS | 7558-79-4 |
---|---|
Molecular Formula | HNa2O4P |
Molecular Weight (g/mol) | 141.96 |
MDL Number | MFCD00003496 |
InChI Key | BNIILDVGGAEEIG-UHFFFAOYSA-L |
Synonym | disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt |
PubChem CID | 24203 |
ChEBI | CHEBI:34683 |
IUPAC Name | disodium;hydrogen phosphate |
SMILES | [Na+].[Na+].OP([O-])([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316778
|
Thermo Scientific Alfa Aesar
013437.30 |
250 g |
25.43 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306778
|
Thermo Scientific Alfa Aesar
013437.A1 |
1 kg |
69.24 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396768
|
Thermo Scientific Alfa Aesar
013437.A7 |
5 kg |
236.18 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium hydrogen phosphate is used in food and water treatment in conjunction with trisodium phosphate. It is used as an emulsifier and buffer in foods and pharmaceuticals, anti-caking additive in powdered products, dietary supplement, scale inhibitor, reagent and buffer in analytical chemistry. It finds application in detergents, cleaning agents, textile, leather auxiliary, fertilizers and galvanoplastics. Further, it is used as a saline laxative to treat constipation. It inhibits the calcium scale formation in water treatment plants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7558-79-4 | |
141.96 | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
24203 | |
disodium;hydrogen phosphate |
HNa2O4P | |
MFCD00003496 | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
CHEBI:34683 | |
[Na+].[Na+].OP([O-])([O-])=O |
Specifications
7558-79-4 | |
Granules | |
HNa2O4P | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
disodium;hydrogen phosphate | |
24203 | |
141.98 | |
Odorless | |
White granular |
White | |
99.0% min. | |
MFCD00003496 | |
Soluble in water. Insoluble in ethanol,methanol and n-octanol. | |
[Na+].[Na+].OP([O-])([O-])=O | |
141.96 | |
CHEBI:34683 | |
ACS | |
Hygroscopic | |
Sodium hydrogen phosphate, anhydrous |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
missing translation for 'einecsNumber' : 231-448-7
missing translation for 'rtecsNumber' : WC4500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only