missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrosulfide hydrate, pure, flakes, Thermo Scientific Chemicals

Product Code. 10461301
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic Bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207683-19-0
Molecular Formula HNaS
Molecular Weight (g/mol) 56.06
MDL Number MFCD00149796
InChI Key HYHCSLBZRBJJCH-UHFFFAOYSA-M
Synonym sodium hydrosulfide hydrate, hnas.h2o, nahs.h2o, sodium sulfanide hydrate, sodium bisulfide monohydrate, sodium hydrosulphide hydrate, sodium hydro sulfide hydrate, sodium hydrosulfide monohydrate, sodium hydrate sulfanide, sodium hydrosulfide hydrate, cp
PubChem CID 16211346
IUPAC Name sodium sulfanide
SMILES [Na+].[SH-]

Specifications

Melting Point 55.0°C
Physical Form Deliquescing Flakes
Quantity 25 g
Linear Formula NaSH·xH2O
Fieser 03,266
Flash Point 90°C
Merck Index 14,8587
Solubility Information Solubility in water: 620g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 56.06
Percent Purity ≥67% min. (NaHS) typically 70 to 72%
Grade Pure
Odor Obnoxious
Packaging Glass bottle
Chemical Name or Material Sodium hydrosulfide hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'