missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 0.1 N standard solution, Thermo Scientific Chemicals

Sodium hydroxide, HNaO, CAS Number-1310-73-2, 7732-18-5, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1L, CHEBI:32145, Beige to White, 0.0995 to 0.1005N (20 deg.C), 1.0000g/mL

35.58 € - 339.00 €

Specifications

Name Note 0.1 N Standard Solution
CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Linear Formula NaOH
Concentration 0.0995 to 0.1005N (20°C)
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10637092
View Documents
Thermo Scientific Acros
124190010
1 L Plastic bottle
35.58 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10348573
View Documents
Thermo Scientific Acros
124190025
2.5 L Plastic Bottle
54.17 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15179173
View Documents
Thermo Scientific Acros
124190250
25 L Plastic Can
339.00 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
Specifications

Specifications

0.1 N Standard Solution
HNaO
0.0995 to 0.1005N (20°C)
01,1083; 05,616; 07,336; 08,460
15, 8761
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Crystalline Powder
1310-73-2, 7732-18-5
NaOH
MFCD00003548
1.0000g/mL
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
[OH-].[Na+]
39.997
CHEBI:32145
Pure
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only