missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hydroxide, 0.1M, Chem-Lab Product Code.: 11973213
compliance-icons

Sodium Hydroxide, 0.1M, Chem-Lab

Product Code. 11973213
25 L, PE
Click to view available options
Quantity:
25 L
Packaging:
PE
This item is not returnable. View return policy

Product Code. 11973213

Brand: Chem Lab CL05.1411.9025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For laboratory use, Titrations, ASTM D-3712, D-4047

Specifications

CAS 1310-73-2
Molecular Formula HNaO
Concentration 4g NaOH / 1L H2O
UN Number UN1824
Density 1.01g/mL
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 25 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Normality 0.1N (±0.0002 at 20°C)
Packaging PE
Chemical Name or Material Sodium hydroxide
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide 0.1 mol/l
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hydroxide, 0.1M, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.