missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 32%, Chem Lab™

Product Code. 11969222
25 L
Click to view available options
Quantity:
25 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

pH >13
Color Colorless
Concentration Sodium hydroxide 32 weight % solution a.r.
Density 1.349g/mL
Solubility Information Soluble in Water
Quantity 25 L
Odor Odorless
Physical Form Liquid
compliance-icons
Product Identifier
  • Sodium hydroxide 32 weight % solution a.r.
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : NaOH/H2O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hydroxide, 32%, Chem Lab™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.