missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 40% w/v aq. soln.

Product Code. p-7050113
Click to view available options
Quantity:
4 L
20 L
Unit Size:
20 L
4 L
This item is not returnable. View return policy

Product Code. 11363767

Brand: Thermo Scientific Alfa Aesar 035639.K7

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1310-73-2 | HNaO | 39.997 g/mol

Sodium hydroxide, 40% w/v aq. solution is used as a chemical reactant or neutralization agent in various industries like chemical, pharmaceutical, textile, pulp and paper, metal and food industries and soap and detergent manufacturing. It is also used in the manufacture of synthetic fabrics such as rayon and also for manufacturing of food products. It is utilized for pH adjustment and formation of in situ sodium methylate in bioethanol and biodiesel processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

UN Number UN1824
Concentration 40% w/v aq. soln.
Solubility Information Miscible with glycerol,ethanol and methanol. Immiscible with acetone and ether.
Formula Weight 40
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Sodium hydroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.