missing translation for 'onlineSavingsMsg'
Learn More
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals Product Code.: 10187850

Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals

Product Code. 10187850
2.5 kg
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
25 kg
This item is not returnable. View return policy

Product Code. 10187850

Brand: Thermo Scientific Acros 424330025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, causticShow MoreShow Less
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color White
Boiling Point 1390.0°C
Melting Point 318.0°C
Assay Percent Range 97+%
Linear Formula NaOH
Fieser 01,1083; 05,616; 07,336; 08,460
Merck Index 15, 8761
Solubility Information Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol,insoluble in acetone
Quantity 2.5 kg
Molecular Weight (g/mol) 40
Formula Weight 40
Percent Purity ≥97%
Grade ACS Reagent
Packaging Plastic Bottle
Vapor Pressure 1mbar at 700°C
Physical Form Pellet
Chemical Name or Material Sodium hydroxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0456762 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog Number424330250,424330010,424330251,424335000,424330100,424330025
Lot NumberA0453016 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number424330250,424330010,424335000,424330251,424330100,424330025
Lot NumberA0466917 Certificate TypeCertificate of Analysis Date21/10/2024 Catalog Number424330250,424330010,424330251,424335000,424330100,424330025
Lot NumberA0466918 Certificate TypeCertificate of Analysis Date21/10/2024 Catalog Number424330250,424330010,424330251,424335000,424330100,424330025
Lot NumberA0466916 Certificate TypeCertificate of Analysis Date21/10/2024 Catalog Number424330250,424330010,424330251,424335000,424330100,424330025
compliance-icons
Product Identifier
  • Sodium hydroxide
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.