missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide, 98%, extra pure, pellets, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Beige to Brown |
Boiling Point | 1390.0°C |
Melting Point | 318.0°C |
Assay Percent Range | 98% |
Linear Formula | NaOH |
Fieser | 01,1083; 05,616; 07,336; 08,460 |
Density | 2.13g/mL |
Merck Index | 15, 8761 |
Solubility Information | Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol, insoluble in acetone |
Quantity | 2.5 kg |
Show More |
Safety and Handling
Product Identifier
- Sodium hydroxide
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction