missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Concentrate, for 1M solution, Honeywell Fluka™

Product Code. 15625250
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

for 10L standard solution, 1 M NaOH (1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
UN Number UN1824
Quantity 6 x 1 Ea.
Formula Weight 40
Vapor Pressure 3 mmHg (37°C)
Chemical Name or Material Sodium hydroxide concentrate, for 10L standard solution, 1 M NaOH (1N)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.