missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, Dilute, EP Grade, Reagecon™
BIOPHARMA

Product Code. 12663623
Click to view available options
Quantity:
1 L
Packaging:
Tamper evident
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured according to Eur. Pharm. Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Sodium Hydroxide Solution, Dilute, EP Grade, Reagecon™ is a ready-to-use reagent with 8.5g sodium hydroxide per 100mL water for use in manual and automated titrations.

Sodium Hydroxide is an inorganic compound consisting of sodium cations Na+ and hydroxide anions OH. It is frequently used a matrix and reagent in a variety of laboratory applications. This solution is prepared by dissolving 8.5g of sodium hydroxide in water and diluted with the water to 100mL.
  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use product eliminates the time and labor of preparing precise reagents and volumetric solutions

Specifications

Color Colorless
CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Concentration 8.5g/100ml
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Soluble in water
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 39.997g/mol
Grade Eu. Ph. Chapter 4(4.1.1)
Packaging Tamper evident
Solvent or Matrix Water
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
compliance-icons
Product Identifier
  • SODIUM HYDROXIDE SOLUTION, DILUTE
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'