missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, extra pure, 33wt.% solution in water, Thermo Scientific Chemicals

Product Code. 11628318
Click to view available options
Quantity:
500 mL
5 L
25 L
Packaging:
Plastic Bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
Boiling Point 120.0°C
Melting Point 8.0°C
Linear Formula NaOH
Fieser 01,1083; 05,616; 07,336; 08,460
Density 1.3600g/mL
Solubility Information Solubility in water: soluble.
Quantity 25 L
Formula Weight 40
Percent Purity 32.5 to 33.5% (total base)
Grade Extra Pure
Packaging Plastic drum
Specific Gravity 1.36
Physical Form Liquid
Chemical Name or Material Sodium hydroxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'