Learn More
Sodium hydroxide (low chloride), ACS, 97.0% min, Thermo Scientific Chemicals
For Use in Chemical Manufacture, Rayon and Cellophane, As a Neutralizing Agent in Petroleum Refining
19.88 € - 590.94 €
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
MDL Number | MFCD00003548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11440320
|
Thermo Scientific Alfa Aesar
013455.88 |
2.5 g | N/A | ||||||
11366818
|
Thermo Scientific Alfa Aesar
013455.22 |
100 g |
43.52 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376818
|
Thermo Scientific Alfa Aesar
013455.36 |
500 g |
50.45 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356818
|
Thermo Scientific Alfa Aesar
013455.A3 |
2 kg |
81.30 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336818
|
Thermo Scientific Alfa Aesar
013455.A9 |
10 kg |
302.51 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346818
|
Thermo Scientific Alfa Aesar
013455.F6 |
2 x 12.5 kg |
590.94 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In chemical manufacture, rayon and cellophane, as a neutralizing agent in petroleum refining, in pulp, paper detergents and textiles, and as a lab reagent. NaOH solutions precipitate most metals as hydroxides from aqueous solutions of the metal saltsSodium hydroxide is widely used in variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as water treatment. It finds application in the base- driven hydrolysis of esters, amides and alkyl halides. It is also used as an industrial cleaning agent, an additive in drilling mud to increase alkalinity in bentonite in petroleum industry and as a powerful degreaser on stainless steel and glass bakeware. It is utilized in the Maxam-Gilbert DNA sequencing technique, chocolate and cocoa processing, soft drink processing, caramel coloring production and for thickening ice cream. It is often used to neutralize acids and prepare sodium salts of reagents. It forms a solution with sodium dodecyl sulfate and plays an important role for the decontamination of mycobacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
1310-73-2 | |
Pellets | |
HNaO | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
ACS | |
White pellets | |
Sodium hydroxide, (low chloride) |
White | |
97.0% min. | |
MFCD00003548 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
Odorless | |
3.5 hPa (3mm Hg) at 800°C |
Safety and Handling
H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
missing translation for 'dotInformation' : Hazard Class: 8, Packing Group: II
missing translation for 'einecsNumber' : 215-185-5
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.