missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide NF, Multi-compendial, J.T. Baker™

Product Code. 15538164
Click to view available options
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Meets BP/ Ph. Eur. Chemical Specifications

Specifications

Boiling Point 1390°C (1013 hPa)
CAS 1310-73-2
Melting Point 323°C
Assay Percent Range 97.0 to 100.5%
Molecular Formula HNaO
MDL Number MFCD00003548
UN Number 1823
Density 2.13 g/cm³ (20°C)
Synonym Caustic soda
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 500 g
SMILES [OH-].[Na+]
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Identification Passes Test
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'