missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 1N standard solution

Product Code. 10784074 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
5 L
25 L
Packaging:
Plastic Bottle
Plastic Can
Plastic drum
Unit Size:
1 L
25 L
5 L
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10784074

Brand: Thermo Scientific Acros 124260050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 1310-73-2 | HNaO | 39.997 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium hydroxide
Color Colorless
CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Linear Formula NaOH
Concentration 0.998 to 1.002N (20°C)
Fieser 01,1083; 05,616; 07,336; 08,460
Density 1.0400g/mL
Merck Index 15, 8761
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and glycerol
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 5 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40
Packaging Plastic drum
Specific Gravity 1.04
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.