missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide, Puriss., meets analytical spec. of BP, NF, Ph. Eur., E524, 98-100.5%, pellets, Honeywell Fluka™
Puriss., meets analytical specification of BP, NF, Ph. Eur., E524, 98-100.5%, pellets
52.13 € - 646.26 €
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
MDL Number | MFCD00003548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15660810
|
Honeywell-Fluka
06203-1KG |
1 kg |
52.13 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15690810
|
Honeywell-Fluka
06203-5KG |
5 kg |
186.67 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15600820
|
Honeywell-Fluka
06203-6X1KG |
6 x 1 kg | N/A | ||||||
15680810
|
Honeywell-Fluka
06203-4X5KG |
4 x 5 kg |
646.26 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15670810
|
Honeywell-Fluka
06203-25KG |
25 kg |
312.09 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
1310-73-2 | |
NaOH | |
1823 | |
Soluble in water, 1260g/L at 20°C | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
98 to 100.5% | |
>1 (vs air) | |
Sodium Hydroxide, Meets analytical specification of Ph. Eur.BP, NF |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40g/mol | |
puriss. | |
3mmHg (37°C), 18mmHg (20°C) |
Safety and Handling
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H290-H314
missing translation for 'einecsNumber' : 215-185-5
missing translation for 'rtecsNumber' : WB4900000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium hydroxide
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title