missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., K ≤0.02%, ≥98%, pellets, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
5 kg
20 kg
4 x 5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Boiling Point | 1,390°C |
Melting Point | 318°C (literature) |
CAS Max % | 100.0000% |
Linear Formula | NaOH |
UN Number | UN1823 |
Density | 2.13 g/cm3 |
Quantity | 4 x 5 kg |
Formula Weight | 40.00 |
Percent Purity | ≥98% |
Grade | ACS Reagent/Reagent Ph. Eur. |
Show More |
Safety and Handling
Product Identifier
- Sodium hydroxide
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'