missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, 0.01M, Honeywell™

Product Code. 15604910 Shop All Honeywell Fluka Products
10 L
Click to view available options
Quantity:
10 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Concentration 0.01M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Quantity 10 L
Vapor Pressure 3 mmHg (37°C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium hydroxide solution, 0.01M, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.