missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution 0.2M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Description
HNaO, CAS Number-1310-73-2, sodium hydroxide, sodium hydroxide solution, sodium hydroxide na oh, sodium hydrate, soda, caustic, white caustic, soda lye, aetznatron, caustic soda, ascarite, 1L, CHEBI:32145, Colorless, 39.99g/mol, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997
Specifications
Specifications
pH | 13 |
Color | Colorless |
Quantity | 5 L |
Formula Weight | 39.99g/mol |
Packaging | Plastic container |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Sodium hydroxide, 0.2 N standard solution
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction