missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis

Product Code. 10498980 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Unit Size:
1 L
10 L
2.5 L
25 L
5 L
This item is not returnable. View return policy

Product Code. 10498980

Brand: Fisher Chemical J/7800/25

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number 3548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH 12
Boiling Point 120°C
Melting Point 8°C
Quantity 25 L
Packaging Plastic container
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.