missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution, ≥20%, Honeywell Fluka™
Click to view available options
Quantity:
1 L
2.5 L
25 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Linear Formula | NaOH |
UN Number | 1824 |
Quantity | 25 L |
Formula Weight | 40g/mol |
Percent Purity | ≥20% |
Vapor Pressure | 3mmHg (37°C) |
Chemical Name or Material | Sodium Hydroxide Solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'