missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution 2M, Pure, Bench Reagent, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
Unit Size:
2.5 L
25 L
Specifications
Specifications
| pH | 14 |
| Color | Colorless |
| Boiling Point | 105°C |
| Melting Point | -10°C |
| Quantity | 25 L |
| Packaging | Plastic container |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction