missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide solution, 0.1M EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12900894 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
This item is not returnable. View return policy

Product Code. 12900894

Brand: Fisher Chemical VL 052 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Bottle
CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Concentration 0.1 M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES [OH-].[Na+]
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
PubChem CID 14798
Quantity 1000 mL
ChEBI CHEBI:32145
Grade Eu. Ph. Reference
Chemical Name or Material Potassium hydroxide solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.