missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, Purum, ≥32%, Honeywell Fluka™
Click to view available options
Quantity:
2.5 L
5 L
35 kg
4 x 2.5 L
4 x 5 L
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Linear Formula | NaOH |
UN Number | 1824 |
Quantity | 5 L |
Formula Weight | 40g/mol |
Percent Purity | ≥32% |
Grade | purum |
Vapor Pressure | 3mmHg (37°C) |
Chemical Name or Material | Sodium Hydroxide Solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'