missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, Volumetric, 0.2 M NaOH (0.2N), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
2 L
Packaging:
Poly Bottle
Unit Size:
2 L
500mL
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Boiling Point | 100°C |
| CAS Min % | ≥99.0000%, ≥0.5000% |
| CAS Max % | >99%, <1.0000% |
| Linear Formula | NaOH |
| Concentration | 0.2 M NaOH (0.2N) |
| UN Number | UN1824 |
| Density | 1.01 g/mL at 25 °C |
| Quantity | 2 L |
| Formula Weight | 40 |
| Grade | Volumetric |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction