missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, Volumetric, 0.5 M NaOH (0.5N), Honeywell Fluka™

Product Code. 15614900 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
5 L
10 L
6 x 1 L
Packaging:
Cubitainer™
Poly Bottle

Volumetric, 0.5 M NaOH (0.5N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Boiling Point >100°C
CAS Min % ≥90.0000%, ≥1.0000%
CAS Max % ≤100.0000%, <2.0000%
Linear Formula NaOH
Concentration 0.5 M NaOH (0.5N)
UN Number UN1824
Density 1.020 g/cm3
Quantity 1 L
Formula Weight 40.00
Grade Volumetric
Packaging Poly Bottle
Chemical Name or Material Sodium hydroxide solution, Volumetric, 0.5 M NaOH (0.5N)
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide; caustic soda
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.