missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hypochlorite Solution, Reagent grade, Available chlorine 4.00 to 4.99%, Honeywell
Click to view available options
Quantity:
25 mL
500 mL
3 L
5 L
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Quantity | 5 L |
Linear Formula | NaClO |
UN Number | 3082 |
Formula Weight | 74.44g/mol |
Grade | Reagent |
Concentration or Composition (by Analyte or Components) | Available chlorine 4.00 to 4.99% |
Vapor Pressure | 17.5mmHg (20°C) |
Density | 1.097g/mL at 25°C |
Chemical Name or Material | Sodium Hypochlorite Solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'