missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite Solution, Reagent grade, Available chlorine 4.00 to 4.99%, Honeywell

Product Code. 15638340
Click to view available options
Quantity:
25 mL
500 mL
3 L
5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, available chlorine 4.00-4.99 %

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Quantity 25 mL
Linear Formula NaClO
UN Number 3082
Formula Weight 74.44g/mol
Grade Reagent
Concentration or Composition (by Analyte or Components) Available chlorine 4.00 to 4.99%
Vapor Pressure 17.5mmHg (20°C)
Density 1.097g/mL at 25°C
Chemical Name or Material Sodium Hypochlorite Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'