missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypophosphite monohydrate, for analysis, Thermo Scientific Chemicals

Product Code. 10411841
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 90.0°C
CAS 10039-56-2
Quantity 100 g
Molecular Formula H4NaO3P
Linear Formula NaH2PO2·H2O
MDL Number MFCD06252379
Synonym sodium hypophosphite hydrate, sodium phosphinate hydrate, sodium phosphenite hydrate, sodium hypophosphite en grade, sodium hydrate hypophosphite, sodium hypophosphite monohydrate 250g, phosphinic acid sodium salt monohydrate, sodium hypophosphite monohydrate, phosphonous acid monosodium salt monohydrate, sodium hypophosphite monohydrate, puriss
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: 33.0 g/100 g ethylene glycol (25°C), 9.7 g/100 g propylene glycol (25°C), freely soluble in glycerol and boiling alcohol, slightly soluble in absolute alcohol, insoluble in ether - soluble in cold
InChI Key PLZNPHDJGFDNRM-UHFFFAOYSA-M
SMILES O.[Na+].[O-][PH2]=O
IUPAC Name sodium hydrate phosphinate
Molecular Weight (g/mol) 105.99
PubChem CID 23708894
Formula Weight 105.99
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Sodium hypophosphite monohydrate
Show More Show Less
Product Identifier
  • Sodium hypophosphite monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'