Learn More
Sodium iodate, 99% min, Thermo Scientific Chemicals
Description
Sodium iodate is used in animal feed, food industry. It is utilized in the manufacture of disinfectants, antiseptics and deodorants. It acts as a significant part in the oxidation of aromatic, aliphatic, and alicyclic thiols. Potassium tetrahydroborate and sodium iodate in acidic aqueous solution react with cadmium to generate chemical vapor, which is useful for atomic fluorescence spectrometry application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | (decomposition) |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 100 g |
UN Number | UN1479 |
Merck Index | 14,8630 |
Solubility Information | Soluble in water and acetone. |
Formula Weight | 197.89 |
Percent Purity | ≤99% |
Sensitivity | Light and moisture sensitive |
Show More |
Safety and Handling
- Sodium iodate
- Danger
- Acute toxicity Category 4
- Oxidising solid Category 2
- Skin sensitiser Category 1
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
- MIXTURE LIST-Contains : Sodium iodate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.