Learn More
Sodium iodide, 99+%, pure, anhydrous, Thermo Scientific Chemicals
34.67 € - 1739.66 €
Chemical Identifiers
CAS | 7681-82-5 |
---|---|
Molecular Formula | INa |
Molecular Weight (g/mol) | 149.89 |
MDL Number | MFCD00003532 |
InChI Key | FVAUCKIRQBBSSJ-UHFFFAOYSA-M |
Synonym | sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech |
PubChem CID | 5238 |
ChEBI | CHEBI:33167 |
IUPAC Name | sodium;iodide |
SMILES | [Na+].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10154640
|
Thermo Scientific Acros
203180050 |
5 g |
34.67 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10206980
|
Thermo Scientific Acros
203182500 |
250 g |
96.68 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10616252
|
Thermo Scientific Acros
203180010 |
1 kg |
276.38 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10689253
|
Thermo Scientific Acros
203180025 |
2.5 kg |
579.89 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15264674
|
Thermo Scientific Acros
203180100 |
10 kg |
1739.66 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7681-82-5 | |
149.89 | |
FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
5238 | |
sodium;iodide |
INa | |
MFCD00003532 | |
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
CHEBI:33167 | |
[Na+].[I-] |
Specifications
1300.0°C | |
7681-82-5 | |
Crystalline Powder | |
INa | |
MFCD00003532 | |
15,8765 | |
Solubility in water: 184 g/100mL (25°C). Other solubilities: soluble in alcohol, glycerol and acetone | |
[Na+].[I-] | |
149.89 | |
CHEBI:33167 | |
≥99% | |
3% max. (1 g, 105°C, 3 hrs) | |
3.66 | |
Sodium iodide, Anhydrous |
661.0°C | |
White | |
99% min. (on dried substance) (Argentometry) | |
NaI | |
01,1087; 02,384; 03,267; 04,456; 06,543; 07,338; 10,365; 11,488; 12,29; 13,43; 14,89; 16,86 | |
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
sodium;iodide | |
5238 | |
149.89 | |
Pure | |
Glass bottle | |
3.6600g/mL |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid releas
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-679-3
- Sodium iodide
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P273-Avoid release to the environment.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.