Learn More
Sodium iodide, ACS, 99.5% min, Thermo Scientific Chemicals
Used to prevent iodine deficiency. Used as analytical reagents, in manufacture of disinfectants, antiseptics, deodorants and medicines, in baking ingredient conditioner. Also used for conversion of alkyl chlorides into alkyl iodides
62.70 € - 215.00 €
Chemical Identifiers
CAS | 7681-82-5 |
---|---|
Molecular Formula | INa |
Molecular Weight (g/mol) | 149.894 |
MDL Number | MFCD00003532 |
InChI Key | FVAUCKIRQBBSSJ-UHFFFAOYSA-M |
Synonym | sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech |
PubChem CID | 5238 |
ChEBI | CHEBI:33167 |
IUPAC Name | sodium;iodide |
SMILES | [Na+].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11379337
|
Thermo Scientific Alfa Aesar
040196.22 |
100 g |
62.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389337
|
Thermo Scientific Alfa Aesar
040196.36 |
500 g |
215.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium iodide commonly is used to prevent iodine deficiency. Used as analytical reagents, in the manufacture of disinfectants, antiseptics, deodorants and medicines. They are used in baking ingredient conditioner and as animal feed. It is used for conversion of an alkyl chlorides into an alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications- Commonly used to prevent iodine deficiency Used as analytical reagents, in manufacture of disinfectants, antiseptics, deodorants and medicines
- Used in baking ingredient conditioner and as animal feed
- Used for conversion of an alkyl chlorides into an alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements
Chemical Identifiers
7681-82-5 | |
149.894 | |
FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
5238 | |
sodium;iodide |
INa | |
MFCD00003532 | |
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
CHEBI:33167 | |
[Na+].[I-] |
Specifications
1300°C to 1304°C | |
7681-82-5 | |
Crystalline Powder | |
INa | |
14,8631 | |
Soluble in water, alcohol, glycerol and acetone. Insoluble in ether. | |
[Na+].[I-] | |
149.894 | |
CHEBI:33167 | |
ACS | |
1.7745 | |
White Powder | |
Sodium iodide |
651°C | |
White | |
99.5% min. | |
MFCD00003532 | |
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
sodium;iodide | |
5238 | |
149.89 | |
Odorless | |
Hygroscopic | |
3.67 g/cm3 at 20°C |
Safety and Handling
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a: Avoid breathing dust/fume/gas/mist/vapours/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Take off contaminated clothing and wash before reuse. Call a POISON CENTER or doctor/physician if you feel unwell. Specific treatment (see label). Store locked up. Store in a well-ventilated place. Keep container tightly closed. Dispose of contents/container in accordance with local/regional/national/international regulations.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 231-679-3
missing translation for 'rtecsNumber' : WB6475000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Hygroscopic and incompatible with strong acids,oxidizing agents. light, air and moisture sensitive.
Recommended Storage : Ambient temperatures
RUO – Research Use Only