missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Iodide, puriss., Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
283.10 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 500 g
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 kg |
Linear Formula | NaI |
UN Number | 3077 |
Formula Weight | 149.89g/mol |
Percent Purity | 99 to 100.5% (calc. to the dried substance) |
Grade | puriss. |
Packaging | Plastic Bottle |
Density | Vapor Density: >1 (vs air) |
Chemical Name or Material | Sodium Iodide, Meets analytical specification of Ph.Eur. BP, USP |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'