Learn More
Sodium isopropoxide, Thermo Scientific Chemicals
58.90 € - 509.00 €
Chemical Identifiers
CAS | 683-60-3 |
---|---|
Molecular Formula | C3H7NaO |
Molecular Weight (g/mol) | 82.078 |
MDL Number | MFCD00061566 |
InChI Key | WBQTXTBONIWRGK-UHFFFAOYSA-N |
Synonym | sodium propan-2-olate, sodium isopropoxide, 2-propanol, sodium salt, isopropoxysodium, iprona, naoipr, 2-propanol, sodium salt 1:1, i-prona, nao-ipr, sodium isopropanolate |
PubChem CID | 10920443 |
IUPAC Name | sodium;propan-2-olate |
SMILES | CC(C)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11483770
|
Thermo Scientific Alfa Aesar
089446.06 |
5 g |
58.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403780
|
Thermo Scientific Alfa Aesar
089446.14 |
25 g |
166.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493770
|
Thermo Scientific Alfa Aesar
089446.22 |
100 g |
509.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium isopropoxide is used in the preparation of ibuprofen intermediate. It is involved in reactions such as polymerization of butadiene. Further, it acts as a catalyst in the manufacturing of acetone and its derivatives. In addition, it is used as a solvent and serves as an antiseptic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
683-60-3 | |
82.078 | |
WBQTXTBONIWRGK-UHFFFAOYSA-N | |
10920443 | |
CC(C)[O-].[Na+] |
C3H7NaO | |
MFCD00061566 | |
sodium propan-2-olate, sodium isopropoxide, 2-propanol, sodium salt, isopropoxysodium, iprona, naoipr, 2-propanol, sodium salt 1:1, i-prona, nao-ipr, sodium isopropanolate | |
sodium;propan-2-olate |
Specifications
683-60-3 | |
C3H7NaO | |
Air and moisture sensitive | |
Soluble in tetrahydrofuran. | |
CC(C)[O-].[Na+] | |
82.078 | |
82.08 | |
Sodium isopropoxide |
70°C to 75°C | |
MFCD00061566 | |
sodium propan-2-olate, sodium isopropoxide, 2-propanol, sodium salt, isopropoxysodium, iprona, naoipr, 2-propanol, sodium salt 1:1, i-prona, nao-ipr, sodium isopropanolate | |
WBQTXTBONIWRGK-UHFFFAOYSA-N | |
sodium;propan-2-olate | |
10920443 | |
Crystalline |
Safety and Handling
P210-P235+P410-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H314-H335-X
missing translation for 'dotInformation' : Hazard Class: 4.2; Packaging Group: II
missing translation for 'einecsNumber' : 211-673-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only