missing translation for 'onlineSavingsMsg'
Learn More

Sodium Meta-Silicate, 9-Hydrate, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15558184
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 13517-24-3
Molecular Formula H18Na2O12Si
Molecular Weight (g/mol) 284.197
InChI Key PHIQPXBZDGYJOG-UHFFFAOYSA-N
Synonym sodium metasilicate nonahydrate, unii-d8d44215lz, sodium silicate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, disodium oxosilanebis olate nonahydrate, 2na.so3i.9h2o, sodium meta-silicate nonahydrate, disodium oxosilanediolate nonahydrate, disodium dioxido oxo silane nonahydrate, sodium oxosilanebis olate-water 1/9
PubChem CID 61639
IUPAC Name disodium;dioxido(oxo)silane;nonahydrate
SMILES O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Quantity 500 g
Formula Weight 284.2
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Chemical Name or Material Sodium Meta-Silicate, 9-Hydrate
compliance-icons compliance-icons
Product Identifier
  • Sodium meta-Silicate, 9-Hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sodium meta-Silicate, 9-Hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.