Learn More
Sodium metabisulfite, 97+%, ACS reagent, Thermo Scientific Chemicals
30.75 € - 84.02 €
Chemical Identifiers
CAS | 7681-57-4 |
---|---|
Molecular Formula | Na2O5S2 |
Molecular Weight (g/mol) | 190.09 |
MDL Number | MFCD00167602 |
InChI Key | LDTLADDKFLAYJA-UHFFFAOYSA-L |
Synonym | sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis |
PubChem CID | 656671 |
IUPAC Name | disodium (sulfinatooxy)sulfinate |
SMILES | [Na+].[Na+].[O-]S(=O)OS([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10361351
|
Thermo Scientific Acros
419580050 |
5 g | Glass Bottle |
30.75 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10444235
|
Thermo Scientific Acros
419582500 |
250 g | Plastic bottle |
40.50 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10301311
|
Thermo Scientific Acros
419580010 |
1 kg | Plastic bottle |
43.82 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10267903
|
Thermo Scientific Acros
419580025 |
2.5 kg | Plastic bottle |
84.02 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
7681-57-4 | |
190.09 | |
LDTLADDKFLAYJA-UHFFFAOYSA-L | |
656671 | |
[Na+].[Na+].[O-]S(=O)OS([O-])=O |
Na2O5S2 | |
MFCD00167602 | |
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis | |
disodium (sulfinatooxy)sulfinate |
Specifications
150.0°C | |
White | |
97% min. (Iodimetry) | |
Na2S2O5 | |
15, 8770 | |
Solubility in water: 540g/L (20°C). Other solubilities: freely soluble in glycerol, slightly soluble in alcohol | |
[Na+].[Na+].[O-]S(=O)OS([O-])=O | |
190.09 | |
190.1 | |
ACS Reagent |
7681-57-4 | |
Fine Crystalline Powder | |
Na2O5S2 | |
MFCD00167602 | |
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis | |
LDTLADDKFLAYJA-UHFFFAOYSA-L | |
disodium (sulfinatooxy)sulfinate | |
656671 | |
≥97% | |
Sodium metabisulfite |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Contact with acids liberates toxic gas.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-673-
- Sodium metabisulfite
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only