missing translation for 'onlineSavingsMsg'
Learn More

Sodium metabisulfite, 97%, Thermo Scientific Chemicals

27.34 € - 143.72 €

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
PubChem CID 656671
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11493587
View Documents
Thermo Scientific Alfa Aesar
A17351.36
500 g
27.34 €
500g
Please to purchase this item. Need a web account? Register with us today!
11403597
View Documents
Thermo Scientific Alfa Aesar
A17351.0E
2500 g
45.43 €
2500g
Please to purchase this item. Need a web account? Register with us today!
11413597
View Documents
Thermo Scientific Alfa Aesar
A17351.0C
10,000 g
143.72 €
10000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium metabisulfite is used as an active ingredient in campden tablets, as a cleaning agent for potable water reverse osmosis membranes, as a preservative and an antioxidant in food. It also acts as a corrosion inhibitor in oil and gas industry, as a bleaching agent in the production of coconut cream, as the source of sulfur dioxide and in the destruction of cyanide in commercial gold cyanidation processes. It is utilized in the precipitation of gold from auric acid as well as in the waste water treatment to remove hexavaent chromium as trivalent chromium by precipitation after reduction. Further, it is used in photography. In addition, it plays as an oxygen scavenger to eliminate the dissolved oxygen in wastewater and in pipes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium metabisulfite is used as an active ingredient in campden tablets, as a cleaning agent for potable water reverse osmosis membranes, as a preservative and an antioxidant in food. It also acts as a corrosion inhibitor in oil and gas industry, as a bleaching agent in the production of coconut cream, as the source of sulfur dioxide and in the destruction of cyanide in commercial gold cyanidation processes. It is utilized in the precipitation of gold from auric acid as well as in the waste water treatment to remove hexavaent chromium as trivalent chromium by precipitation after reduction. Further, it is used in photography. In addition, it plays as an oxygen scavenger to eliminate the dissolved oxygen in wastewater and in pipes.

Solubility
Soluble in water, ethanol and glycerol.

Notes
Moisture and air sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications

Chemical Identifiers

7681-57-4
190.09
LDTLADDKFLAYJA-UHFFFAOYSA-L
656671
[Na+].[Na+].[O-]S(=O)OS([O-])=O
Na2O5S2
MFCD00167602
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
disodium (sulfinatooxy)sulfinate

Specifications

>300°C
3.5 to 5.0
Na2S2O5
14,8638
Soluble in water,ethanol and glycerol.
[Na+].[Na+].[O-]S(=O)OS([O-])=O
190.09
190.1
Like Rotten Eggs
1.48
7681-57-4
Na2O5S2
MFCD00167602
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
LDTLADDKFLAYJA-UHFFFAOYSA-L
disodium (sulfinatooxy)sulfinate
656671
97%
Air and moisture sensitive
Sodium metabisulfite
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c

H302-H318

missing translation for 'einecsNumber' : 231-673-0

missing translation for 'rtecsNumber' : UX8225000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • Sodium metabisulfite

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.

Precautionary Statement
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : sodium metabisulphate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only