Learn More
Sodium metatungstate, 2.82 density solution in water, stabilized, Thermo Scientific Chemicals
168.84 € - 455.27 €
Chemical Identifiers
CAS | 12141-67-2 |
---|---|
Molecular Formula | H2Na6O40W12 |
MDL Number | MFCD00150663 |
Synonym | Sodium polytungstate |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10603702
|
Thermo Scientific Acros
304100500 |
50 mL | Glass bottle |
168.84 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10693502
|
Thermo Scientific Acros
304102500 |
250 mL | Glass bottle |
455.27 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12141-67-2 | |
H2Na6O40W12 | |
MFCD00150663 | |
Sodium polytungstate |
Specifications
Green to Yellow | |
H2Na6O40W12 | |
MFCD00150663 | |
Sodium polytungstate | |
2986.13 | |
2.82 | |
Sodium metatungstate, 2.82 Density Solution in Water, Stabilized |
12141-67-2 | |
Na6[H2W12O40] | |
2.8200g/mL | |
Solubility in water: >1000g/L water (20°C). | |
≥67% (WO3) | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'tsca' : TSCA
- Sodium metatungstate, 2.82 density solution in water, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only