missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium metavanadate, typically 96%, V 38% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
250 g
1 kg
Description
Sodium metavanadate is used as an analytical reagent and mordant. It is also found to be useful in the photography industry and the pharmaceutical industry. In addition to this, it is used in the making of vanadium alloy and vanadium catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 630°C |
Color | White |
Physical Form | Chunks and/or Powder |
Quantity | 25 g |
Assay Percent Range | Typically 96% |
Solubility Information | Soluble in water. |
IUPAC Name | sodium;oxido(dioxo)vanadium |
Formula Weight | 121.93 |
Odor | Odorless |
Appearance | Off-white to light orange powder |
Show More |
Safety and Handling
Product Identifier
- Sodium metavanadate
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- H301-Toxic if swallowed.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H361-Suspected of damaging fertility or the unborn child.
- H372-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Sodium metavanadate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'