missing translation for 'onlineSavingsMsg'
Learn More
Sodium methanesulfonate, 98%, Thermo Scientific Chemicals Product Code.: 11410587

Sodium methanesulfonate, 98%, Thermo Scientific Chemicals

Product Code. 11410587
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11410587

Brand: Thermo Scientific Alfa Aesar B22439.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium methanesulfonate is used as a non-steroidal anti-inflammatory drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium methanesulfonate is used as a non-steroidal anti-inflammatory drug.

Solubility
Soluble in water, alcohol, ether, hexane, benzene, methylcyclopentane, toluene, o-chlorotoluene and ethyl disulfide.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-57-4
Molecular Formula CH3NaO3S
Molecular Weight (g/mol) 118.082
MDL Number MFCD00064389
InChI Key KKVTYAVXTDIPAP-UHFFFAOYSA-M
Synonym sodium methanesulfonate, methanesulfonic acid sodium salt, methanesulfonic acid, sodium salt, sodium methanesulphonate, sodium mesylate, methylsulfonic acid sodium salt, 75-75-2 parent, sodium methyl sulfonate, methanesulfonic acid, sodium salt 1:1, sodium methylsulfonateShow MoreShow Less
PubChem CID 638112
IUPAC Name sodium;methanesulfonate
SMILES CS(=O)(=O)[O-].[Na+]

Specifications

Odor Odorless
Linear Formula CH3SO3Na
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol,ether,hexane,benzene,methylcyclopentane,toluene,o-chlorotoluene and ethyl disulfide.
Formula Weight 118.09
Percent Purity 98%
Chemical Name or Material Sodium methanesulfonate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253315 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog NumberB22439.06,B22439.14
Lot Number10193822 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22439.06,B22439.14
Lot Number10116438 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22439.06,B22439.14
Lot Number10205774 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22439.06,B22439.14
Lot Number10182594 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22439.06,B22439.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium methanesulfonate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.