missing translation for 'onlineSavingsMsg'
Learn More
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals Product Code.: 10357853

Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals

Product Code. 10357853
10 L
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
25 L
This item is not returnable. View return policy

Product Code. 10357853

Brand: Thermo Scientific Acros 168600100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-41-4, 67-56-1
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.02
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Melting Point 1.0°C to 5.0°C
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 93.0°C
Flash Point 33°C
Packaging Plastic drum
Linear Formula CH3ONa
Refractive Index 1.3701
Quantity 10 L
Beilstein 01,28
Fieser 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
Merck Index 15,8775
Specific Gravity 0.97
Solubility Information Solubility in water: reacts. Other solubilities: soluble in methanol
Viscosity 68 mPa.s (20°C)
Formula Weight 54.02
Grade Pure
Physical Form Liquid
Chemical Name or Material Sodium methoxide, 30 wt% Solution in Methanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467897 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number168600025,168600100,168600010,168600250,168600251
Lot NumberA0458271 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number168600025,168600100,168600010,168600250,168600251
Lot NumberA0458272 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number168600025,168600100,168600010,168600250,168600251
Lot NumberA0465329 Certificate TypeCertificate of Analysis Date09/08/2024 Catalog Number168600025,168600100,168600010,168600250,168600251
Lot NumberA0462364 Certificate TypeCertificate of Analysis Date01/05/2024 Catalog Number168600025,168600100,168600010,168600250,168600251

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.